This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310455_m1
See other GUCY1B2 GE Assays ›
Gene Symbol:
Gucy1b2
Gene Name:
guanylate cyclase 1, soluble, beta 2
Gene Aliases:
B930025B03, GC-SB2
Chromosome Location:
Chr.14: 62373023 - 62457562 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310455_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gucy1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204340.1 NP_001191269.1 11 - 12 1666 61
NM_172810.3 NP_766398.1 11 - 12 1711 61
GenBank mRNA AK047129.1 - 11 - 12 1712 61
AY236513.1 - 9 - 10 1385 61
BC137868.1 - 11 - 12 1663 8860887 61
BC144988.1 - 11 - 12 1660 9053536 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002206.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gucy1b2

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Process(es)

guanylate cyclase activity
ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017062.mm.1 NM_001204340.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017062.mm.1 NM_001204340.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017062.mm.1 NM_172810.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017062.mm.1 NM_172810.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017062.mm.1 TC1400002206.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017063.mm.1 NM_001204340.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017063.mm.1 NM_001204340.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017063.mm.1 NM_172810.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017063.mm.1 NM_172810.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017063.mm.1 TC1400002206.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0