This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308912_m1
See other D930007P13RIK GE Assays ›
Gene Symbol:
D930007P13Rik
Gene Name:
Riken cDNA D930007P13 gene
Gene Aliases:
-
Chromosome Location:
Chr.15: 103123070 - 103146828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308912_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D930007P13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045743.1 6 - 7 1104 89
GenBank mRNA AK052984.1 - 6 - 7 1105 89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB522487.2 - 3 - 4 419 Chr. 15 - 103123074 - 103127036 [-] on Build GRCm38 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
30528
Gm30528 -59.3kb Sense Chr. 15 - 103026805 - 103063780 on NCBI Build GRCm38
Downstream single-strand selective monofunctional uracil DNA glycosylase Smug1 1200013B09Rik,
A930006H09Rik,
C85220
27.4kb Sense Chr. 15 - 103153290 - 103168895 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002308.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039404 AK052984.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123310 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039404 AK052984.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08488869 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123310 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08488829 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122291 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123310 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08488829 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12277159 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502263 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502283 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502393 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12277144 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502266 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502267 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12277148 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502277 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502394 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502280 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502281 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502272 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502270 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502261 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30502278 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500019096.mm.1 NR_045743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019096.mm.1 NR_045743.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019096.mm.1 TC1500002308.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 874676 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0