This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308907_m1
See other D930007P13RIK GE Assays ›
Gene Symbol:
D930007P13Rik
Gene Name:
Riken cDNA D930007P13 gene
Gene Aliases:
-
Chromosome Location:
Chr.15: 103123070 - 103146828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308907_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D930007P13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045743.1 1 - 2 519 90
GenBank mRNA AK052984.1 - 1 - 2 520 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB662554.1 - 1 - 2 509 Chr. 15 - 103144008 - 103146818 [-] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
30528
Gm30528 -81.7kb Sense Chr. 15 - 103026805 - 103063780 on NCBI Build GRCm38
Downstream single-strand selective monofunctional uracil DNA glycosylase Smug1 1200013B09Rik,
A930006H09Rik,
C85220
6.5kb Sense Chr. 15 - 103153290 - 103168895 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002308.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500019101.mm.1 NR_045743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019101.mm.1 NR_045743.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019101.mm.1 TC1500002308.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019102.mm.1 NR_045743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019102.mm.1 NR_045743.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019102.mm.1 TC1500002308.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header