This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01307400_m1
See other NEK9 GE Assays ›
Gene Symbol:
Nek9
Gene Name:
NIMA (never in mitosis gene a)-related expressed kinase 9
Gene Aliases:
C130021H08Rik
Chromosome Location:
Chr.12: 85299514 - 85339362 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01307400_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nek9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145138.2 NP_660120.2 20 - 21 2763 79
GenBank mRNA AJ489828.1 - 20 - 21 2738 79
AK173323.1 - 20 - 21 2747 79
BC024926.1 - 2 - 3 350 79
BC117971.2 - 20 - 21 2704 40098375 79
BC117972.2 - 20 - 21 2703 40098378 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nek9

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cell cycle
mitotic nuclear division
phosphorylation
cell division
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0