This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01306990_g1
See other IGF2BP1 GE Assays ›
Gene Symbol:
Igf2bp1
Gene Name:
insulin-like growth factor 2 mRNA binding protein 1
Gene Aliases:
AL024068, AW549074, CRD-BP, Crdbp, D030026A21Rik, D11Moh40e, D11Moh45, IMP-1, Neilsen, ZBP-1, Zbp1, mir-3063
Chromosome Location:
Chr.11: 95957163 - 96005944 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01306990_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igf2bp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009951.4 NP_034081.1 3 - 4 593 145
GenBank mRNA AF061569.1 - 3 - 4 412 145
AK013940.1 - 3 - 4 560 145
AK044850.1 - 3 - 4 498 145
BC051679.1 - 3 - 4 576 30008106 145

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igf2bp1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

RNA localization
regulation of translation
transport
regulation of mRNA stability involved in response to stress
negative regulation of translation
pallium cell proliferation in forebrain
mRNA transport
CRD-mediated mRNA stabilization
neuronal stem cell population maintenance
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100033044.mm.1 NM_009951.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033044.mm.1 NM_009951.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100033044.mm.1 TC1100003604.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0