This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305456_g1
See other PPP1R37 GE Assays ›
Gene Symbol:
Ppp1r37
Gene Name:
protein phosphatase 1, regulatory subunit 37
Gene Aliases:
Gm158, Lrrc68
Chromosome Location:
Chr.7: 19530967 - 19563071 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305456_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r37
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199149.3 NP_954600.1 6 - 7 1100 87
GenBank mRNA AK050946.1 - 6 - 7 1101 87
AK160935.1 - 6 - 7 1101 87
BC070402.1 - 6 - 7 948 5696223 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002487.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r37

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
protein phosphatase inhibitor activity
phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700021891.mm.1 NM_199149.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700021891.mm.1 NM_199149.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700021891.mm.1 TC0700002487.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0