This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305300_m1
See other SPATS2 GE Assays ›
Gene Symbol:
Spats2
Gene Name:
spermatogenesis associated, serine-rich 2
Gene Aliases:
2700012F11Rik, 59kDa, Scr59, p59
Chromosome Location:
Chr.15: 99125926 - 99213215 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305300_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spats2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139140.1 NP_631879.1 10 - 11 1322 85
GenBank mRNA AB079132.1 - 10 - 11 1322 85
AK159335.1 - 10 - 11 1417 85
AK167788.1 - 9 - 10 1000 85
AK171266.1 - 9 - 10 1078 85
BC023432.1 - 8 - 9 972 5029821 85
BC072579.1 - 10 - 11 1589 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spats2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0