This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304524_m1
See other IPO11 GE Assays ›
Gene Symbol:
Ipo11
Gene Name:
importin 11
Gene Aliases:
1700081H05Rik, 2510001A17Rik, AI314624, AW555235, E330021B14Rik, Ranbp11
Chromosome Location:
Chr.13: 106794439 - 106936959 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304524_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ipo11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029665.3 NP_083941.2 7 - 8 864 93
GenBank mRNA AK044487.1 - 7 - 8 896 93
AK087793.1 - 7 - 8 897 93
BC031900.1 - 7 - 8 800 5364208 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002617.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ipo11

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

ribosomal protein import into nucleus
transport
intracellular protein transport
protein transport
protein binding
Ran GTPase binding
protein transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017818.mm.1 NM_029665.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017818.mm.1 NM_029665.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017818.mm.1 TC1300002617.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017817.mm.1 NM_029665.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017817.mm.1 NM_029665.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017817.mm.1 TC1300002617.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017816.mm.1 NM_029665.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017816.mm.1 NM_029665.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017816.mm.1 TC1300002617.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0