This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304024_m1
See other FYB GE Assays ›
Gene Symbol:
Fyb
Gene Name:
FYN binding protein
Gene Aliases:
ADAP, B630013F22Rik, FYB-120/130, Slap130, p120/130, p120/p130
Chromosome Location:
Chr.15: 6522797 - 6665608 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304024_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fyb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278269.1 NP_001265198.1 7 - 8 1731 82
NM_011815.5 NP_035945.1 7 - 8 1731 82
GenBank mRNA AF001863.1 - 7 - 8 1707 82
AF061744.1 - 7 - 8 1707 82
AK038225.1 - 7 - 8 1759 82
AK137907.1 - 7 - 8 1687 82
BC141352.1 - 7 - 8 1672 9055964 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000037.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fyb

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

mast cell activation
protein localization to plasma membrane
protein binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500000304.mm.1 NM_001278269.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500000304.mm.1 NM_011815.5 Refseq
Affymetrix Mouse Clariom S Array PSR1500000304.mm.1 TC1500000037.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500000305.mm.1 NM_001278269.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500000305.mm.1 NM_011815.5 Refseq
Affymetrix Mouse Clariom S Array PSR1500000305.mm.1 TC1500000037.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0