This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303915_m1
See other L3MBTL2 GE Assays ›
Gene Symbol:
L3mbtl2
Gene Name:
l(3)mbt-like 2 (Drosophila)
Gene Aliases:
4732493N06Rik, M4mbt
Chromosome Location:
Chr.15: 81663889 - 81688315 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303915_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: L3mbtl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289711.1 NP_001276640.1 15 - 16 2002 75
NM_001289712.1 NP_001276641.1 15 - 16 2002 75
NM_145993.5 NP_666105.2 15 - 16 2002 75
NR_110362.1 15 - 16 2325 75
GenBank mRNA AK029115.1 - 15 - 16 1947 75
AK036510.1 - 15 - 16 1955 75
AK132012.1 - 15 - 16 1954 75
AK141810.1 - 14 - 15 3135 75
AK146632.1 - 15 - 16 1958 75
AK156236.1 - 15 - 16 1957 75
AY237000.1 - 15 - 16 1935 75
AY237001.1 - 15 - 16 1935 75
AY237002.1 - 15 - 16 2258 75
BC023676.1 - 4 - 5 537 75
BC023933.1 - 15 - 16 1943 5325131 75
BC030864.1 - 15 - 16 1905 4211489 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000726.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene L3mbtl2

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
chromatin modification
zinc ion binding
methylated histone binding
histone binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500005640.mm.1 NM_001289711.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005640.mm.1 NM_001289712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005640.mm.1 NM_145993.5 Refseq
Affymetrix Mouse Clariom S Array PSR1500005640.mm.1 NR_110362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005640.mm.1 TC1500000726.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0