This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303722_m1
See other ARHGAP8 GE Assays ›
Gene Symbol:
Arhgap8
Gene Name:
Rho GTPase activating protein 8
Gene Aliases:
3110043J09Rik
Chromosome Location:
Chr.15: 84720052 - 84772857 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303722_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164627.1 NP_001158099.1 4 - 5 288 63
NM_001164628.1 NP_001158100.1 3 - 4 280 63
NM_001205334.1 NP_001192263.1 4 - 5 308 63
NM_028455.4 NP_082731.2 3 - 4 300 63
GenBank mRNA AK040142.1 - 3 - 4 250 63
AK086816.1 - 3 - 4 254 63
BC005563.1 - 2 - 3 243 3965919 63
BC010306.1 - 3 - 4 270 63
BY723460.1 - 3 - 4 274 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000793.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap8

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_001164627.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_001164627.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_001164628.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_001164628.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_001205334.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_001205334.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_028455.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 NM_028455.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500006251.mm.1 TC1500000793.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0