This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302592_m1
See other RAD51B GE Assays ›
Gene Symbol:
Rad51b
Gene Name:
RAD51 homolog B
Gene Aliases:
AI553500, R51H2, Rad51l1, mREC2
Chromosome Location:
Chr.12: 79297282 - 79852895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302592_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rad51b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252562.1 NP_001239491.1 4 - 5 371 105
NM_009014.3 NP_033040.2 4 - 5 389 105
GenBank mRNA AK015771.1 - 4 - 5 371 105
AK032913.1 - 4 - 5 390 105
U92068.1 - 3 - 4 331 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rad51b

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
in utero embryonic development
blastocyst growth
DNA repair
DNA recombination
cellular response to DNA damage stimulus
positive regulation of cell proliferation
positive regulation of G2/M transition of mitotic cell cycle
somite development
nucleotide binding
four-way junction DNA binding
DNA binding
double-stranded DNA binding
single-stranded DNA binding
ATP binding
DNA-dependent ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header