This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302591_m1
See other RAD51B GE Assays ›
Gene Symbol:
Rad51b
Gene Name:
RAD51 homolog B
Gene Aliases:
AI553500, R51H2, Rad51l1, mREC2
Chromosome Location:
Chr.12: 79297282 - 79852895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302591_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rad51b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252562.1 NP_001239491.1 2 - 3 137 92
NM_009014.3 NP_033040.2 2 - 3 155 92
GenBank mRNA AK015771.1 - 2 - 3 137 92
AK032913.1 - 2 - 3 156 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000719.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rad51b

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
in utero embryonic development
blastocyst growth
DNA repair
DNA recombination
cellular response to DNA damage stimulus
positive regulation of cell proliferation
positive regulation of G2/M transition of mitotic cell cycle
somite development
nucleotide binding
four-way junction DNA binding
DNA binding
double-stranded DNA binding
single-stranded DNA binding
ATP binding
DNA-dependent ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200005132.mm.1 NM_001252562.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005132.mm.1 NM_001252562.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005132.mm.1 NM_009014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005132.mm.1 NM_009014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200005132.mm.1 TC1200000719.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200005131.mm.1 NM_001252562.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005131.mm.1 NM_001252562.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005131.mm.1 NM_009014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005131.mm.1 NM_009014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200005131.mm.1 TC1200000719.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200005129.mm.1 NM_001252562.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005129.mm.1 NM_001252562.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005129.mm.1 TC1200000719.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0