This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302413_m1
See other DDX52 GE Assays ›
Gene Symbol:
Ddx52
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 52
Gene Aliases:
2700029C06Rik, ROK1
Chromosome Location:
Chr.11: 83942090 - 83963088 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302413_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030096.2 NP_084372.2 3 - 4 508 128
GenBank mRNA AK080767.1 - 3 - 4 446 128
AK133545.1 - 3 - 4 479 128
AK141200.1 - 3 - 4 482 128
AK166789.1 - 3 - 4 434 128
BC029094.1 - 3 - 4 420 3662249 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001272.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx52

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
biological_process
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100011927.mm.1 NM_030096.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100011927.mm.1 NM_030096.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100011927.mm.1 TC1100001272.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0