This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302120_m1
See other DENND2C GE Assays ›
Gene Symbol:
Dennd2c
Gene Name:
DENN/MADD domain containing 2C
Gene Aliases:
A930010I20Rik
Chromosome Location:
Chr.3: 103102654 - 103169738 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302120_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dennd2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177857.1 NP_808525.2 9 - 10 1926 58
GenBank mRNA AK044389.1 - 8 - 9 2302 58
AK137637.1 - 4 - 5 533 58
AK172232.1 - 5 - 6 648 58
BC046440.1 - 4 - 5 394 58
BC060666.1 - 9 - 10 1926 6841982 58
BC066182.1 - 2 - 3 172 58

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dennd2c

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
Rab guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0