This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01301134_g1
See other JMJD8 GE Assays ›
Gene Symbol:
Jmjd8
Gene Name:
jumonji domain containing 8
Gene Aliases:
2610003J06Rik
Chromosome Location:
Chr.17: 25829043 - 25831843 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01301134_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Jmjd8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028101.4 NP_082377.2 5 - 6 432 68
GenBank mRNA AK011308.1 - 5 - 6 429 68
AK148079.1 - 5 - 6 410 68
AK155810.1 - 5 - 6 434 68
AK172073.1 - 5 - 6 429 68
BC010800.1 - 5 - 6 394 4235072 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000391.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Jmjd8

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700003308.mm.1 NM_028101.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003308.mm.1 NM_028101.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700003308.mm.1 TC1700000391.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700003307.mm.1 NM_028101.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003307.mm.1 NM_028101.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700003307.mm.1 TC1700000391.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0