This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299430_m1
See other SCUBE1 GE Assays ›
Gene Symbol:
Scube1
Gene Name:
signal peptide, CUB domain, EGF-like 1
Gene Aliases:
7330410C13Rik, A630023E24Rik
Chromosome Location:
Chr.15: 83602583 - 83725049 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299430_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Scube1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271472.1 NP_001258401.1 20 - 21 2807 62
NM_001271473.1 NP_001258402.1 17 - 18 2474 62
NM_022723.3 NP_073560.2 19 - 20 2717 62
GenBank mRNA AF276425.1 - 19 - 20 2584 62
AK041590.1 - 19 - 20 2700 62
AK083722.1 - 3 - 4 666 62
BC066066.1 - 20 - 21 2765 6850999 62
BC080278.1 - 17 - 18 2442 30354974 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001980.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Scube1

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

protein homooligomerization
calcium ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500015663.mm.1 NM_001271472.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015663.mm.1 NM_001271473.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015663.mm.1 NM_022723.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500015663.mm.1 TC1500001980.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0