This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299097_m1
See other TTLL8 GE Assays ›
Gene Symbol:
Ttll8
Gene Name:
tubulin tyrosine ligase-like family, member 8
Gene Aliases:
1700019P01Rik, 4933401B17
Chromosome Location:
Chr.15: 88889778 - 88955641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299097_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttll8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172818.3 NP_766406.2 8 - 9 1411 82
GenBank mRNA AK029745.1 - 8 - 9 1407 82
AK030151.1 - 8 - 9 1404 82
AM690752.1 - 8 - 9 1068 82
BC116293.1 - 8 - 9 1052 40094952 82
BC116294.1 - 8 - 9 1047 40094955 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002032.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ttll8

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein polyglycylation
cilium assembly
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016110.mm.1 NM_172818.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016110.mm.1 NM_172818.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500016110.mm.1 TC1500002032.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header