This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01298480_m1
See other SLC38A9 GE Assays ›
Gene Symbol:
Slc38a9
Gene Name:
solute carrier family 38, member 9
Gene Aliases:
4833412L08Rik, 6720411P22Rik, 9130023D20Rik, 9430067K09Rik, A730092C09
Chromosome Location:
Chr.13: 112660691 - 112738752 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01298480_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc38a9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178746.4 NP_848861.1 2 - 3 250 114
GenBank mRNA AK043402.1 - 2 - 3 238 114
AK044735.1 - 2 - 3 240 114
BC052361.1 - 2 - 3 250 6705789 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001266.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc38a9

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
positive regulation of TOR signaling
cellular response to amino acid stimulus
amino acid transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300008027.mm.1 NM_178746.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008027.mm.1 NM_178746.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300008027.mm.1 TC1300001266.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0