This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296760_mH
See other NLRP4F GE Assays ›
Gene Symbol:
Nlrp4f
Gene Name:
NLR family, pyrin domain containing 4F
Gene Aliases:
C330026N02Rik, C86045, Nalp-kappa, Nalp4f
Chromosome Location:
Chr.13: 65177111 - 65205716 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296760_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nlrp4f
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175290.4 NP_780499.3 3 - 4 1970 122
GenBank mRNA AK049352.1 - 1 - 2 373 122
AY596201.1 - 4 - 5 2080 122
BC098479.1 - 1 - 2 392 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002211.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nlrp4f

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
biological_process
nucleotide binding
molecular_function
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300014320.mm.1 NM_175290.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014320.mm.1 NM_175290.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300014320.mm.1 TC1300002211.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014321.mm.1 NM_175290.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014321.mm.1 NM_175290.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300014321.mm.1 TC1300002211.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014322.mm.1 NM_175290.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014322.mm.1 NM_175290.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300014322.mm.1 TC1300002211.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014319.mm.1 NM_175290.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014319.mm.1 NM_175290.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300014319.mm.1 TC1300002211.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014318.mm.1 NM_175290.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014318.mm.1 NM_175290.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300014318.mm.1 TC1300002211.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header