This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295847_m1
See other PHLPP1 GE Assays ›
Gene Symbol:
Phlpp1
Gene Name:
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases:
AI836256, Phlpp, Plekhe1, SCOP, mKIAA0606
Chromosome Location:
Chr.1: 106171869 - 106394245 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295847_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phlpp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133821.3 NP_598582.3 13 - 14 3450 119
GenBank mRNA AB093251.1 - 13 - 14 2209 119
BC024670.1 - 1 - 2 78 119
BC059254.1 - 13 - 14 2110 6406203 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001018.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phlpp1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
protein dephosphorylation
apoptotic process
entrainment of circadian clock
negative regulation of protein kinase B signaling
negative regulation of protein kinase C signaling
catalytic activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100008446.mm.1 NM_133821.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008446.mm.1 NM_133821.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100008446.mm.1 TC0100001018.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100008445.mm.1 NM_133821.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008445.mm.1 NM_133821.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100008445.mm.1 TC0100001018.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0