This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295432_m1
See other RNF2 GE Assays ›
Gene Symbol:
Rnf2
Gene Name:
ring finger protein 2
Gene Aliases:
AI326319, AI450156, AU019207, Ring1B, dinG
Chromosome Location:
Chr.1: 151469406 - 151500923 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295432_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011277.2 NP_035407.1 1 - 2 107 157
GenBank mRNA AK012358.2 - 1 - 2 108 157
AK013124.1 - 1 - 2 98 157
AK090066.1 - 1 - 2 91 157
AK145767.1 - 1 - 2 114 157
BC020122.1 - 1 - 2 80 4021046 157

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK012358.2, AK013124.1, AK090066.1, AK145767.1, BC020122.1, NM_011277.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnf2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
gastrulation with mouth forming second
transcription, DNA-templated
regulation of transcription, DNA-templated
anterior/posterior axis specification
histone ubiquitination
histone H2A monoubiquitination
histone H2A-K119 monoubiquitination
negative regulation of sequence-specific DNA binding transcription factor activity
chromatin binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
RING-like zinc finger domain binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100026431.mm.1 NM_011277.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026431.mm.1 NM_011277.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100026431.mm.1 TC0100003254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100026433.mm.1 NM_011277.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026433.mm.1 NM_011277.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100026433.mm.1 TC0100003254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100026434.mm.1 NM_011277.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026434.mm.1 NM_011277.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100026434.mm.1 TC0100003254.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 922710 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0