This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295113_s1
See other PPP1R2-PS9 GE Assays ›
Gene Symbol:
Ppp1r2-ps9
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 2, pseudogene 9
Gene Aliases:
4930403L05Rik
Chromosome Location:
Chr.X: 15110586 - 15111489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295113_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r2-ps9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033171.1 1 627 96
GenBank mRNA AK015065.1 - 1 632 96
AK015311.1 - 1 604 96
BC100448.1 - 1 604 30918995 96
BC132484.1 - 1 576 40130761 96
BC132486.1 - 1 576 40130763 96

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase inhibitor phosphatase modulator

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00012049.mm.1 NR_033171.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012049.mm.1 TC0X00001883.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header