This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01294823_m1
See other 1700017G19RIK GE Assays ›
Gene Symbol:
1700017G19Rik
Gene Name:
RIKEN cDNA 1700017G19 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 40504865 - 40522912 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01294823_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700017G19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040445.1 5 - 6 1295 70
GenBank mRNA AK006064.1 - 5 - 6 1296 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream FAT tumor suppressor homolog 4 (Drosophila) Fat4 6030410K14Rik,
9430004M15
-1.5mb Anti-sense Chr. 3 - 38886487 - 39011987 on NCBI Build GRCm38
Downstream inturned planar cell polarity effector homolog (Drosophila) Intu 9230116I04Rik,
9430087H23Rik,
Pdzd6,
Pdzk6,
mKIAA1284
146.7kb Anti-sense Chr. 3 - 40653702 - 40704774 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137246 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386802 AK006064.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386802 AK006064.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177713 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139498 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177713 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139498 - -
Whole Mouse Genome G4122A A_51_P386802 AK006064.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P386802 AK006064.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101099 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177713 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139498 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080449 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080377 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02441545 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03108930 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080470 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080516 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02381801 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080513 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02883265 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080447 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02790591 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080374 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02383467 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080476 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080455 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080439 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080389 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080383 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02790588 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080485 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02343804 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080484 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080376 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080395 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080518 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080469 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02355413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080371 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03096910 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02664104 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03108929 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080406 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080423 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03010580 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02441553 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080375 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080429 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03030059 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02790585 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080444 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080388 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080381 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080454 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080458 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02523856 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080372 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080385 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080393 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080452 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02381808 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02790583 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02790582 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080522 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03108933 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02511621 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02883270 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080369 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080479 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080441 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22080392 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1429645_PM_at - -
Mouse Genome 430 2.0 Array 1429645_PM_at - -
Mouse Genome 430 2.0 Array 1429645_at - -
Mouse Expression Set 430 1429645_at - -
Mouse Expression Set 430 1429645_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429645_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429645_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 453725 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101780193 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0