This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01294820_m1
See other 1700017G19RIK GE Assays ›
Gene Symbol:
1700017G19Rik
Gene Name:
RIKEN cDNA 1700017G19 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 40504865 - 40522912 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01294820_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700017G19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040445.1 2 - 3 437 61
GenBank mRNA AK006064.1 - 2 - 3 438 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream FAT tumor suppressor homolog 4 (Drosophila) Fat4 6030410K14Rik,
9430004M15
-1.5mb Anti-sense Chr. 3 - 38886487 - 39011987 on NCBI Build GRCm38
Downstream inturned planar cell polarity effector homolog (Drosophila) Intu 9230116I04Rik,
9430087H23Rik,
Pdzd6,
Pdzk6,
mKIAA1284
132.2kb Anti-sense Chr. 3 - 40653702 - 40704774 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001898.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300014588.mm.1 NR_040445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300014588.mm.1 NR_040445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300014588.mm.1 TC0300001898.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0