This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01293018_m1
See other PRKAR2B GE Assays ›
Gene Symbol:
Prkar2b
Gene Name:
protein kinase, cAMP dependent regulatory, type II beta
Gene Aliases:
AI451071, AW061005, PKARIIbeta, Pkarb2, RII(beta)
Chromosome Location:
Chr.12: 31958450 - 32061343 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01293018_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkar2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011158.3 NP_035288.2 5 - 6 777 75
GenBank mRNA AK011647.1 - 5 - 6 735 75
AK041013.1 - 5 - 6 751 75
AK081805.1 - 5 - 6 554 75
AK135621.1 - 5 - 6 779 75
AK138963.1 - 5 - 6 743 75
BC033267.1 - 1 - 2 65 75
BC048710.1 - 5 - 6 733 6506827 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001602.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkar2b

Panther Classification:

Molecular Function -

protein-binding activity modulator kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
fatty acid metabolic process
learning
regulation of protein kinase activity
negative regulation of cAMP-dependent protein kinase activity
nucleotide binding
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein kinase binding
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200011158.mm.1 NM_011158.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011158.mm.1 NM_011158.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200011158.mm.1 TC1200001602.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0