Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100010949.mm.1 |
NM_001159866.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010949.mm.1 |
NM_001159866.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010949.mm.1 |
NM_019562.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010949.mm.1 |
NM_019562.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010949.mm.1 |
TC0100001317.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010950.mm.1 |
NM_001159866.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010950.mm.1 |
NM_001159866.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010950.mm.1 |
NM_019562.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010950.mm.1 |
NM_019562.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010950.mm.1 |
TC0100001317.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010951.mm.1 |
NM_001159866.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010951.mm.1 |
NM_001159866.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010951.mm.1 |
NM_019562.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010951.mm.1 |
NM_019562.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010951.mm.1 |
TC0100001317.mm.1 |
MTA TC |