This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292672_m1
See other TTC1 GE Assays ›
Gene Symbol:
Ttc1
Gene Name:
tetratricopeptide repeat domain 1
Gene Aliases:
4833412C19Rik, C79328, TPR1
Chromosome Location:
Chr.11: 43730006 - 43747973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292672_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133795.1 NP_598556.1 3 - 4 480 92
GenBank mRNA AK077715.1 - 3 - 4 493 92
AK166431.1 - 3 - 4 487 92
AK170032.1 - 3 - 4 490 92
AK170230.1 - 3 - 4 490 92
BC010236.1 - 3 - 4 480 3581904 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002574.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ttc1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100023780.mm.1 NM_133795.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100023780.mm.1 NM_133795.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100023780.mm.1 TC1100002574.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0