This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292331_m1
See other SEPT2 GE Assays ›
Gene Symbol:
Sept2
Gene Name:
septin 2
Gene Aliases:
AW208991, Nedd-5, Nedd5, mKIAA0158
Chromosome Location:
Chr.1: 93478967 - 93509733 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292331_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sept2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010891.2 NP_035021.1 1 - 2 75 73
GenBank mRNA AK028072.1 - 1 - 2 57 73
AK146616.1 - 1 - 2 72 73
AK151591.1 - 1 - 2 59 73
AK171331.1 - 1 - 2 54 73
CJ075082.1 - 1 - 2 98 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK028072.1, AK146616.1, AK151591.1, AK171331.1, NM_010891.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sept2

Panther Classification:

Molecular Function -

cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
regulation of L-glutamate transport
cell cycle
mitotic nuclear division
smoothened signaling pathway
neuron projection development
regulation of protein localization
cilium assembly
regulation of catalytic activity
protein polymerization
cell division
nucleotide binding
GTPase activity
protein binding
GTP binding
enzyme regulator activity
protein complex scaffold
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0