This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292257_m1
See other TOR1AIP1 GE Assays ›
Gene Symbol:
Tor1aip1
Gene Name:
torsin A interacting protein 1
Gene Aliases:
LAP1, Lap1B
Chromosome Location:
Chr.1: 156004599 - 156036480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292257_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tor1aip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160018.1 NP_001153490.1 7 - 8 1064 66
NM_001160019.1 NP_001153491.1 7 - 8 478 66
GenBank mRNA AB251963.1 - 8 - 9 1061 66
AK149727.1 - 7 - 8 1255 66
AK152751.1 - 7 - 8 498 66
BQ891636.1 - 7 - 8 659 66
CA465752.1 - 7 - 8 481 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003313.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of ATPase activity
protein localization to nucleus
nuclear membrane organization
ATPase activator activity
protein binding
lamin binding
cytoskeletal protein binding
ATPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100027045.mm.1 NM_001160018.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027045.mm.1 NM_001160018.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027045.mm.1 NM_001160019.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027045.mm.1 NM_001160019.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027045.mm.1 TC0100003313.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0