This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292254_m1
See other TOR1AIP1 GE Assays ›
Gene Symbol:
Tor1aip1
Gene Name:
torsin A interacting protein 1
Gene Aliases:
LAP1, Lap1B
Chromosome Location:
Chr.1: 156004599 - 156036480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292254_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tor1aip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160018.1 NP_001153490.1 4 - 5 905 122
NM_001160019.1 NP_001153491.1 4 - 5 319 122
NM_144791.2 NP_659040.2 4 - 5 905 122
GenBank mRNA AB251963.1 - 4 - 5 845 122
AK149727.1 - 4 - 5 825 122
AK152751.1 - 4 - 5 339 122
BC010841.1 - 4 - 5 720 3493883 122
BQ891636.1 - 4 - 5 500 122
CA465752.1 - 4 - 5 322 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003313.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tor1aip1

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of ATPase activity
protein localization to nucleus
nuclear membrane organization
ATPase activator activity
protein binding
lamin binding
cytoskeletal protein binding
ATPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 NM_001160018.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 NM_001160018.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 NM_001160019.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 NM_001160019.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 NM_144791.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 NM_144791.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100027051.mm.1 TC0100003313.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 NM_001160018.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 NM_001160018.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 NM_001160019.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 NM_001160019.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 NM_144791.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 NM_144791.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100027053.mm.1 TC0100003313.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0