This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291790_m1
See other 4930533P14RIK GE Assays ›
Gene Symbol:
4930533P14Rik
Gene Name:
RIKEN cDNA 4930533P14 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 96612057 - 96662573 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291790_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930533P14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040478.1 2 - 3 308 106
GenBank mRNA AK015960.1 - 2 - 3 309 106
BY715526.1 - 2 - 3 306 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 29601 Gm29601 -55.3kb Sense Chr. 1 - 96543281 - 96556729 on NCBI Build GRCm38
Downstream solute carrier organic anion transporter family,
member 4C1
Slco4c1 9530051F04,
C330017E21Rik,
OATP-H,
OATP-M1,
OATP4C1,
OATPX,
PRO2176,
SLC21A20,
oatp-R
203.6kb Sense Chr. 1 - 96818784 - 96872171 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002855.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827443 NM_001014761.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540579 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773112 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443331 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428341 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189676 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715610 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693679 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634662 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620105 AK016181.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620105 BC025039.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262842 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175347 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6832 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7914 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844527 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427447 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447469 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85404 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38878 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28482 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04198589 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04198637 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04198565 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119466 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04198589 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04198637 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023103.mm.1 NR_040478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023103.mm.1 NR_040478.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023103.mm.1 TC0100002855.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 856330 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106400195 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0