This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291789_m1
See other 4930533P14RIK GE Assays ›
Gene Symbol:
4930533P14Rik
Gene Name:
RIKEN cDNA 4930533P14 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 96612057 - 96662573 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291789_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930533P14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040478.1 1 - 2 157 69
GenBank mRNA AK015960.1 - 1 - 2 158 69
BY715526.1 - 1 - 2 155 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 29601 Gm29601 -58.3kb Sense Chr. 1 - 96543281 - 96556729 on NCBI Build GRCm38
Downstream solute carrier organic anion transporter family,
member 4C1
Slco4c1 9530051F04,
C330017E21Rik,
OATP-H,
OATP-M1,
OATP4C1,
OATPX,
PRO2176,
SLC21A20,
oatp-R
156.2kb Sense Chr. 1 - 96818784 - 96872171 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002855.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023104.mm.1 NR_040478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023104.mm.1 NR_040478.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023104.mm.1 TC0100002855.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100023105.mm.1 NR_040478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023105.mm.1 NR_040478.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023105.mm.1 TC0100002855.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0