This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291592_m1
See other A830008E24RIK GE Assays ›
Gene Symbol:
A830008E24Rik
Gene Name:
RIKEN cDNA A830008E24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291592_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A830008E24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK043564.1 - 2 - 3 260 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579741 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848101 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188690 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710948 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915825 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344859 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276961 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835677 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467569 AK090063.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522729 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49969 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908626 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268444 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81859 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310067 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505269 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93893 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514200 AK006360.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377365 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93151 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404732 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97586 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651839 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25252 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63400 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426966 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535841 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167268 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104269 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3296 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3887 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132890 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04313257 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00220735 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00220719 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04313111 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04313141 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04313229 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111254 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00220719 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04313257 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00321446 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730325 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730168 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730098 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00022572 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00302681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730352 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730329 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730165 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133207 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00412756 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730376 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730345 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730333 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00205049 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730358 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00228510 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730159 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730163 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730190 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730389 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730195 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00436207 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00818778 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730100 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730245 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730380 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730091 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00022575 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730334 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730347 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730308 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730193 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730189 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730335 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730175 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730387 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730162 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00856813 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730408 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730354 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730384 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00228512 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00313820 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00030668 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00431908 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00441250 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00429552 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730390 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133209 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730105 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730158 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133204 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00022574 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730336 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00313818 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730177 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730197 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00616562 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730164 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730092 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730340 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730355 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730404 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730370 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00818772 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730382 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133210 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730393 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730090 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730361 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00856821 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730094 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00156290 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730156 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730331 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730357 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730364 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730103 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730167 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00631351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00789738 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730406 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730096 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730306 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00990718 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730339 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133206 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00818780 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730087 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00818776 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730187 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730305 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00135831 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730366 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730161 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730323 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730341 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730372 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133211 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01133213 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730377 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730350 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20730410 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0