This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291244_m1
See other EDIL3 GE Assays ›
Gene Symbol:
Edil3
Gene Name:
EGF-like repeats and discoidin I-like domains 3
Gene Aliases:
Del-1, Del1
Chromosome Location:
Chr.13: 88821472 - 89323225 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291244_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Edil3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037987.3 NP_001033076.1 5 - 6 977 75
NM_010103.4 NP_034233.1 4 - 5 947 75
GenBank mRNA AF031524.1 - 5 - 6 1085 75
AF031525.1 - 5 - 6 1016 75
AK036117.1 - 5 - 6 938 75
AK046885.1 - 5 - 6 978 75
AK081003.1 - 4 - 5 779 75
AK135220.1 - 5 - 6 941 75
AK139093.1 - 5 - 6 939 75
AK141907.1 - 5 - 6 961 75
AK157810.1 - 5 - 6 962 75
BC056386.1 - 5 - 6 877 6854497 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000998.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Edil3

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
multicellular organism development
positive regulation of cell-substrate adhesion
integrin binding
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006420.mm.1 NM_001037987.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006420.mm.1 NM_001037987.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006420.mm.1 NM_010103.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006420.mm.1 NM_010103.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006420.mm.1 TC1300000998.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0