This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291241_m1
See other EDIL3 GE Assays ›
Gene Symbol:
Edil3
Gene Name:
EGF-like repeats and discoidin I-like domains 3
Gene Aliases:
Del-1, Del1
Chromosome Location:
Chr.13: 88821472 - 89323225 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291241_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Edil3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037987.3 NP_001033076.1 1 - 2 580 94
NM_010103.4 NP_034233.1 1 - 2 580 94
GenBank mRNA AF031524.1 - 1 - 2 688 94
AF031525.1 - 1 - 2 619 94
AK036117.1 - 1 - 2 541 94
AK044978.1 - 1 - 2 439 94
AK045088.1 - 1 - 2 396 94
AK045839.1 - 1 - 2 497 94
AK046885.1 - 1 - 2 581 94
AK081003.1 - 1 - 2 412 94
AK084215.1 - 1 - 2 435 94
AK135220.1 - 1 - 2 544 94
AK139093.1 - 1 - 2 542 94
AK157810.1 - 1 - 2 565 94
BC056386.1 - 1 - 2 480 6854497 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000998.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Edil3

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
multicellular organism development
positive regulation of cell-substrate adhesion
integrin binding
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006404.mm.1 NM_001037987.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006404.mm.1 NM_001037987.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006404.mm.1 NM_010103.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006404.mm.1 NM_010103.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006404.mm.1 TC1300000998.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006413.mm.1 NM_001037987.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006413.mm.1 NM_001037987.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006413.mm.1 NM_010103.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006413.mm.1 NM_010103.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006413.mm.1 TC1300000998.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006407.mm.1 NM_001037987.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006407.mm.1 NM_001037987.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006407.mm.1 NM_010103.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006407.mm.1 NM_010103.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006407.mm.1 TC1300000998.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006401.mm.1 NM_001037987.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006401.mm.1 NM_001037987.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006401.mm.1 NM_010103.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006401.mm.1 NM_010103.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006401.mm.1 TC1300000998.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 320803 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0