This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291114_m1
See other PLK4 GE Assays ›
Gene Symbol:
Plk4
Gene Name:
polo-like kinase 4
Gene Aliases:
1700028H20, AI385771, Sak, Stk18
Chromosome Location:
Chr.3: 40799951 - 40816883 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291114_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plk4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011495.2 NP_035625.2 2 265
NM_173169.2 NP_775261.2 2 265
NR_045576.1 2 265
GenBank mRNA AK006827.1 - 2 265
AK045082.1 - 2 265
AK137080.1 - 2 265
AK137471.1 - 2 265
AK164465.1 - 2 265
BC026785.1 - 2 5150173 265
BC057940.1 - 2 5323295 265
L29479.1 - 2 265
L29480.1 - 2 265

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plk4

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
centriole replication
phosphorylation
positive regulation of centriole replication
trophoblast giant cell differentiation
de novo centriole assembly
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm