This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291030_m1
See other DIS3L2 GE Assays ›
Gene Symbol:
Dis3l2
Gene Name:
DIS3 mitotic control homolog (S. cerevisiae)-like 2
Gene Aliases:
4930429A22Rik, 8030493P09Rik
Chromosome Location:
Chr.1: 86703779 - 87050097 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291030_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dis3l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172157.1 NP_001165628.1 8 - 9 896 84
NM_153530.2 NP_705758.1 7 - 8 854 84
GenBank mRNA AK029652.1 - 8 - 9 910 84
AK031180.1 - 8 - 9 879 84
AK040613.1 - 9 - 10 951 84
BC036177.1 - 7 - 8 815 5004341 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000794.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dis3l2

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, exonucleolytic
rRNA processing
mRNA catabolic process
cell cycle
mitotic nuclear division
negative regulation of cell proliferation
miRNA catabolic process
rRNA catabolic process
stem cell population maintenance
nuclear-transcribed mRNA catabolic process, exonucleolytic, 3'-5'
cell division
mitotic sister chromatid separation
RNA phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
ribonuclease activity
poly(U) RNA binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100006682.mm.1 NM_001172157.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006682.mm.1 NM_001172157.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006682.mm.1 NM_153530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006682.mm.1 NM_153530.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100006682.mm.1 TC0100000794.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header