This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290698_m1
See other MPZL1 GE Assays ›
Gene Symbol:
Mpzl1
Gene Name:
myelin protein zero-like 1
Gene Aliases:
1110007A10Rik, PZR
Chromosome Location:
Chr.1: 165592181 - 165634558 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290698_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpzl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083897.1 NP_001077366.1 4 - 5 757 67
GenBank mRNA AK090278.1 - 4 - 5 743 67
AK169385.1 - 4 - 5 747 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003449.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

myelin protein

Gene Ontology Categories:

Function(s)

signal transduction

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100028268.mm.1 NM_001083897.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028268.mm.1 NM_001083897.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028268.mm.1 TC0100003449.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0