This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290103_m1
See other EPB4.1L5 GE Assays ›
Gene Symbol:
Epb4.1l5
Gene Name:
erythrocyte protein band 4.1 like 5
Gene Aliases:
1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
Chromosome Location:
Chr.1: 119545032 - 119649136 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290103_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1l5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113416.1 NP_001106887.1 11 - 12 1088 153
NM_145506.4 NP_663481.2 11 - 12 1088 153
GenBank mRNA AK034676.1 - 11 - 12 1010 153
AK037024.1 - 11 - 12 1044 153
AK040354.1 - 11 - 12 1056 153
AK044959.1 - 11 - 12 1026 153
AK044996.1 - 11 - 12 1034 153
AK047523.1 - 11 - 12 1015 153
AK079301.1 - 11 - 12 1039 153
AK136125.1 - 11 - 12 1043 153
AK161141.1 - 11 - 12 1039 153
AK167449.1 - 11 - 12 1088 153
AK173198.1 - 11 - 12 1022 153
BC003937.1 - 11 - 12 1073 3486543 153
BC011476.1 - 11 - 12 1022 4014009 153

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002950.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epb4.1l5

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis involved in differentiation
in utero embryonic development
somitogenesis
epithelial to mesenchymal transition
neural plate morphogenesis
positive regulation of cell-matrix adhesion
epithelial cell morphogenesis
apical constriction
substrate-dependent cell migration, cell attachment to substrate
ectoderm development
endoderm development
mesoderm development
mesoderm migration involved in gastrulation
unidimensional cell growth
posttranscriptional regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-cell adhesion
actin cytoskeleton organization
actomyosin structure organization
negative regulation of protein binding
positive regulation of protein binding
somite rostral/caudal axis specification
axial mesoderm development
axial mesoderm morphogenesis
paraxial mesoderm development
embryonic foregut morphogenesis
positive regulation of focal adhesion assembly
regulation of establishment of protein localization
left/right axis specification
cellular response to transforming growth factor beta stimulus
protein binding
cytoskeletal protein binding
protein domain specific binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023711.mm.1 NM_001113416.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023711.mm.1 NM_001113416.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023711.mm.1 NM_145506.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023711.mm.1 NM_145506.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100023711.mm.1 TC0100002950.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header