This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01289631_m1
See other SPG20 GE Assays ›
Gene Symbol:
Spg20
Gene Name:
spastic paraplegia 20, spartin (Troyer syndrome) homolog (human)
Gene Aliases:
AI840044, C79168, SPARTIN, TAHCCP1, mKIAA0610
Chromosome Location:
Chr.3: 55112074 - 55137332 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01289631_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spg20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144987.1 NP_001138459.1 2 - 3 1133 102
NM_001144988.1 NP_001138460.1 2 - 3 917 102
NM_144895.2 NP_659144.1 2 - 3 917 102
GenBank mRNA AK031545.1 - 1 - 2 472 102
AK035081.1 - 1 - 2 776 102
AK036569.1 - 1 - 2 240 102
AK087388.1 - 2 - 3 885 102
AK089946.1 - 2 - 3 918 102
AK129171.1 - 2 - 3 896 102
AK147760.1 - 2 - 3 1135 102
AK159951.1 - 2 - 3 918 102
BC022921.1 - 2 - 3 885 4220506 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK087388.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spg20

Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
negative regulation of BMP signaling pathway
lipid particle organization
negative regulation of collateral sprouting in absence of injury
neuromuscular process
regulation of mitochondrial membrane potential
adipose tissue development
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300002941.mm.1 NM_001144987.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002941.mm.1 NM_001144987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002941.mm.1 TC0300000416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300002942.mm.1 NM_001144987.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002942.mm.1 NM_001144987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002942.mm.1 TC0300000416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 NM_001144987.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 NM_001144987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 NM_001144988.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 NM_001144988.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 NM_144895.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 NM_144895.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300002948.mm.1 TC0300000416.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0