This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288470_g1
See other MAPKAPK2 GE Assays ›
Gene Symbol:
Mapkapk2
Gene Name:
MAP kinase-activated protein kinase 2
Gene Aliases:
AA960234, MAPKAP-K2, MK-2, MK2, Rps6kc1
Chromosome Location:
Chr.1: 131053700 - 131097543 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288470_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapkapk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008551.1 NP_032577.1 6 - 7 748 69
GenBank mRNA AK208763.1 - 1 - 2 63 69
AK211802.1 - 1 - 2 63 69
AK214789.1 - 1 - 2 63 69
AK215327.1 - 1 - 2 63 69
AK217708.1 - 1 - 2 63 69
AK219589.1 - 1 - 2 63 69
AK219616.1 - 1 - 2 63 69
BC024559.1 - 2 - 3 117 69
BC063064.1 - 6 - 7 748 5687007 69
X76850.1 - 6 - 7 727 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003051.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mapkapk2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

toll-like receptor signaling pathway
protein phosphorylation
response to stress
inflammatory response
cellular response to DNA damage stimulus
phosphorylation
peptidyl-serine phosphorylation
G2 DNA damage checkpoint
response to lipopolysaccharide
regulation of interleukin-6 production
regulation of tumor necrosis factor production
response to cytokine
cellular response to vascular endothelial growth factor stimulus
p38MAPK cascade
positive regulation of tumor necrosis factor biosynthetic process
macropinocytosis
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
mRNA stabilization
inner ear development
3'-UTR-mediated mRNA stabilization
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
signal transducer activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
mitogen-activated protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100024488.mm.1 NM_008551.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024488.mm.1 TC0100003051.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0