This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288151_m1
See other MAOA GE Assays ›
Gene Symbol:
Maoa
Gene Name:
monoamine oxidase A
Gene Aliases:
1110061B18Rik, AA407771
Chromosome Location:
Chr.X: 16619698 - 16687812 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288151_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Maoa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173740.3 NP_776101.3 13 - 14 1509 91
GenBank mRNA AK004337.1 - 13 - 14 1357 91
AK078642.1 - 6 - 7 496 91
AK146612.1 - 13 - 14 1510 91
AK155052.1 - 13 - 14 1372 91
AK164457.1 - 13 - 14 1346 91
BC029100.1 - 13 - 14 1379 3481702 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000181.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Maoa

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

catecholamine metabolic process
neurotransmitter catabolic process
dopamine catabolic process
serotonin metabolic process
phenylethylamine metabolic process
oxidation-reduction process
protein binding
primary amine oxidase activity
oxidoreductase activity
flavin adenine dinucleotide binding
serotonin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00001150.mm.1 NM_173740.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001150.mm.1 NM_173740.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001150.mm.1 TC0X00000181.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0