This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01286095_m1
See other ILKAP GE Assays ›
Gene Symbol:
Ilkap
Gene Name:
integrin-linked kinase-associated serine/threonine phosphatase 2C
Gene Aliases:
0710007A14Rik, 1600009O09Rik, PP2C-DELTA
Chromosome Location:
Chr.1: 91373846 - 91398841 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01286095_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ilkap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023343.2 NP_075832.1 7 - 8 702 65
GenBank mRNA AF117831.1 - 2 - 3 156 65
AK003007.1 - 6 - 7 1675 65
AK005408.1 - 7 - 8 725 65
BC011427.1 - 7 - 8 672 65
BC026953.1 - 7 - 8 702 5133980 65
BC027439.1 - 7 - 8 591 4164301 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002798.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ilkap

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
regulation of nuclear cell cycle DNA replication
catalytic activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100022638.mm.1 NM_023343.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100022638.mm.1 TC0100002798.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0