This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01284705_m1
See other PROSER1 GE Assays ›
Gene Symbol:
Proser1
Gene Name:
proline and serine rich 1
Gene Aliases:
2810046L04Rik, 9330161F11
Chromosome Location:
Chr.3: 53463817 - 53481755 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01284705_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Proser1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173382.1 NP_775558.2 7 - 8 1024 76
GenBank mRNA AK034174.1 - 7 - 8 1175 76
AK038404.1 - 7 - 8 1024 76
AK087909.1 - 7 - 8 1045 76
BC086674.1 - 7 - 8 581 30653602 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK038404.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Proser1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300002745.mm.1 NM_173382.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002745.mm.1 NM_173382.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002745.mm.1 TC0300000396.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0