This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282835_m1
See other SLC30A5 GE Assays ›
Gene Symbol:
Slc30a5
Gene Name:
solute carrier family 30 (zinc transporter), member 5
Gene Aliases:
1810010K08Rik, AF233321, ZNT5, ZTL1, ZnT-5, Zntl1
Chromosome Location:
Chr.13: 100802645 - 100833563 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282835_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc30a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022885.2 NP_075023.2 11 - 12 1685 137
GenBank mRNA AF233321.1 - 4 - 5 704 137
AF461761.1 - 11 - 12 1584 137
AK007424.1 - 2 - 3 205 137
AK051183.1 - 11 - 12 1685 137
AK088004.1 - 10 - 11 1429 137
AK090176.1 - 11 - 12 1594 137
AK166006.1 - 11 - 12 1495 137
BC029033.1 - 8 - 9 1127 137
BC029217.1 - 4 - 5 537 137
BC033452.1 - 11 - 12 1584 3156622 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002544.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc30a5

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
cobalt ion transport
zinc II ion transport
cellular zinc ion homeostasis
response to zinc ion
transmembrane transport
zinc II ion transmembrane transport
zinc ion transmembrane transporter activity
cation transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017366.mm.1 NM_022885.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017366.mm.1 NM_022885.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017366.mm.1 TC1300002544.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0