This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282775_m1
See other PIK3R1 GE Assays ›
Gene Symbol:
Pik3r1
Gene Name:
phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha)
Gene Aliases:
PI3K, p50alpha, p55alpha, p85alpha
Chromosome Location:
Chr.13: 101680761 - 101768217 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282775_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pik3r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077495.2 NP_001070963.1 2 - 3 955 128
GenBank mRNA BB647657.1 - 1 - 2 172 128
BC026146.1 - 2 - 3 936 3979333 128
M60651.1 - 2 - 3 890 128
U50413.1 - 2 - 3 908 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002553.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

cellular glucose homeostasis
positive regulation of protein phosphorylation
negative regulation of cell-matrix adhesion
glucose metabolic process
protein phosphorylation
transport
negative regulation of cell adhesion
signal transduction
insulin receptor signaling pathway
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of heart rate
positive regulation of gene expression
negative regulation of muscle cell apoptotic process
phosphatidylinositol 3-kinase signaling
protein transport
negative regulation of cell-cell adhesion
B cell differentiation
positive regulation of cell migration
positive regulation of tumor necrosis factor production
response to insulin
cellular response to insulin stimulus
positive regulation of RNA splicing
cellular response to UV
response to endoplasmic reticulum stress
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of transcription factor import into nucleus
negative regulation of apoptotic process
regulation of phosphatidylinositol 3-kinase activity
positive regulation of myoblast differentiation
negative regulation of osteoclast differentiation
negative regulation of blood pressure
negative regulation of proteolysis
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol phosphorylation
insulin-like growth factor receptor signaling pathway
negative regulation of smooth muscle cell proliferation
protein stabilization
response to glucocorticoid
regulation of stress fiber assembly
NFAT protein import into nucleus
response to cAMP
growth hormone receptor signaling pathway
regulation of establishment of protein localization to plasma membrane
positive regulation of endoplasmic reticulum unfolded protein response
receptor binding
insulin receptor binding
insulin-like growth factor receptor binding
platelet-derived growth factor receptor binding
neurotrophin TRKA receptor binding
protein binding
calmodulin binding
protein C-terminus binding
transcription factor binding
1-phosphatidylinositol-3-kinase activity
protein kinase binding
protein phosphatase binding
protein domain specific binding
estrogen receptor binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
phosphatidylinositol 3-kinase regulator activity
phosphatidylinositol 3-kinase regulatory subunit binding
ErbB-3 class receptor binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding
1-phosphatidylinositol-3-kinase regulator activity
protein heterodimerization activity
ATPase binding
phosphoprotein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0