This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282560_m1
See other POLK GE Assays ›
Gene Symbol:
Polk
Gene Name:
polymerase (DNA directed), kappa
Gene Aliases:
Dinb1
Chromosome Location:
Chr.13: 96480689 - 96542648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282560_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012048.2 NP_036178.1 11 - 12 1537 97
GenBank mRNA AB027563.1 - 11 - 12 1525 97
AF163571.1 - 11 - 12 1537 97
AY769919.1 - 10 - 11 1351 97
AY769920.1 - 10 - 11 1174 97
AY769921.1 - 9 - 10 1111 97
AY769922.1 - 9 - 10 1111 97
AY769923.1 - 7 - 8 911 97
AY769924.1 - 7 - 8 819 97
AY769925.1 - 10 - 11 1351 97
AY769926.1 - 10 - 11 1351 97
AY769927.1 - 9 - 10 1111 97
BC052820.1 - 8 - 9 1202 30057338 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002471.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polk

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
nucleotide-excision repair, DNA gap filling
cellular response to DNA damage stimulus
DNA biosynthetic process
DNA binding
damaged DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300016544.mm.1 NM_012048.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300016544.mm.1 TC1300002471.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header