This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282370_m1
See other DMGDH GE Assays ›
Gene Symbol:
Dmgdh
Gene Name:
dimethylglycine dehydrogenase precursor
Gene Aliases:
1200014D15Rik, AI787269
Chromosome Location:
Chr.13: 93674436 - 93752823 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282370_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dmgdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028772.3 NP_083048.1 1 - 2 104 80
GenBank mRNA AK004755.1 - 1 - 2 105 80
BC089599.1 - 1 - 2 107 30118948 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001051.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dmgdh

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glycine metabolic process
tetrahydrofolate interconversion
oxidation-reduction process
folic acid binding
oxidoreductase activity
poly(A) RNA binding
dimethylglycine dehydrogenase activity
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006700.mm.1 NM_028772.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006700.mm.1 NM_028772.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006700.mm.1 TC1300001051.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006701.mm.1 NM_028772.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006701.mm.1 NM_028772.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006701.mm.1 TC1300001051.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0