This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01281503_m1
See other POLH GE Assays ›
Gene Symbol:
Polh
Gene Name:
polymerase (DNA directed), eta (RAD 30 related)
Gene Aliases:
RAD30A, XPV
Chromosome Location:
Chr.17: 46171355 - 46202683 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01281503_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030715.3 NP_109640.1 2 - 3 369 82
GenBank mRNA AB027128.1 - 2 - 3 395 82
AK036296.1 - 2 - 3 370 82
BC049159.1 - 2 - 3 340 82
BC128366.1 - 2 - 3 366 40111620 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC128366.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polh

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA synthesis involved in DNA repair
DNA replication
DNA repair
pyrimidine dimer repair
postreplication repair
cellular response to DNA damage stimulus
response to UV-C
cellular response to UV-C
DNA biosynthetic process
DNA binding
damaged DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700020345.mm.1 NM_030715.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700020345.mm.1 NM_030715.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700020345.mm.1 TC1700002167.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header